missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triphenyltetrazolium chloride, 100.2%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210312625

 View more versions of this product

Catalog No. ICN10312625

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2, 3, 5-Triphenyltetrazolium chloride
White
C19H15ClN4
25 g
PKDBCJSWQUOKDO-UHFFFAOYSA-M
2,3,5-triphenyltetrazol-2-ium;chloride
9283
334.8
298-96-4
1.002
MFCD00011963
2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
334.81
CHEBI:78019
Crystalline Powder

Chemical Identifiers

298-96-4
334.81
PKDBCJSWQUOKDO-UHFFFAOYSA-M
9283
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
C19H15ClN4
MFCD00011963
2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride
CHEBI:78019
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions