missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%
Description
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.
Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∽250°C (decomposition) |
| Assay Percent Range | 98% |
| Quantity | 50 g |
| Beilstein | 3923356 |
| Sensitivity | Hygroscopic; Light sensitive |
| Merck Index | 14,9744 |
| Solubility Information | Soluble in water,ethanol and acetone. Insoluble in ether. |
| Formula Weight | 334.81 |
| Percent Purity | 98% |
| Chemical Name or Material | 2, 3, 5-Triphenyl-2H-tetrazolium chloride |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 206-071-6 |
| RTECSNumber | XF8100000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only