missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trimethylpyrazine 98.0+%, TCI America™
$38.51 - $96.26
Chemical Identifiers
| CAS | 14667-55-1 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00006145 |
| InChI Key | IAEGWXHKWJGQAZ-UHFFFAOYSA-N |
| Synonym | trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl |
| PubChem CID | 26808 |
| IUPAC Name | 2,3,5-trimethylpyrazine |
| SMILES | CC1=CN=C(C(=N1)C)C |
Chemical Identifiers
| 14667-55-1 | |
| 122.171 | |
| IAEGWXHKWJGQAZ-UHFFFAOYSA-N | |
| 26808 | |
| CC1=CN=C(C(=N1)C)C |
| C7H10N2 | |
| MFCD00006145 | |
| trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl | |
| 2,3,5-trimethylpyrazine |
Specifications
| 14667-55-1 | |
| 172°C | |
| MFCD00006145 | |
| trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl | |
| CC1=CN=C(C(=N1)C)C | |
| 122.171 | |
| 122.17 | |
| Liquid |
| Yellow | |
| C7H10N2 | |
| 5 g | |
| IAEGWXHKWJGQAZ-UHFFFAOYSA-N | |
| 2,3,5-trimethylpyrazine | |
| 26808 | |
| ≥98.0% (GC,T) | |
| 2,3,5-Trimethylpyrazine |
Safety and Handling
RTECSNumber : UQ3907000
TSCA : Yes