missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylpyrazine 98.0+%, TCI America™

Catalog No. T09425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09425G 5 g
T094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09425G Supplier TCI America Supplier No. T09425G

Chemical Identifiers

CAS 14667-55-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006145
InChI Key IAEGWXHKWJGQAZ-UHFFFAOYSA-N
Synonym trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
PubChem CID 26808
IUPAC Name 2,3,5-trimethylpyrazine
SMILES CC1=CN=C(C(=N1)C)C

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylpyrazine
RTECSNumber UQ3907000
TSCA Yes