missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol 98.0+%, TCI America™

Supplier:  TCI America T0485500G

 View more versions of this product

Catalog No. T0485500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3,5-Trimethylphenol
93°C
233°C
MFCD00002228
2430
OGRAOKJKVGDSFR-UHFFFAOYSA-N
2,3,5-trimethylphenol
12769
136.19
Crystalline Powder
697-82-5
White-Yellow
C9H12O
500 g
isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
CC1=CC(=C(C(=C1)O)C)C
136.194
CHEBI:38570
≥98.0% (GC)

Chemical Identifiers

697-82-5
136.194
OGRAOKJKVGDSFR-UHFFFAOYSA-N
12769
2,3,5-trimethylphenol
C9H12O
MFCD00002228
isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
CHEBI:38570
CC1=CC(=C(C(=C1)O)C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0540&(4)-0057&(9)-0899

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions