missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triméthylphénol 98,0+%, TCI America™

Numéro de catalogue. T048525G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T048525G 25 g
T0485500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T048525G Fournisseur TCI America Code fournisseur T048525G

Identifiants chimiques

CAS 697-82-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002228
InChI Key OGRAOKJKVGDSFR-UHFFFAOYSA-N
Synonym isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
PubChem CID 12769
ChEBI CHEBI:38570
IUPAC Name 2,3,5-triméthylphénol
SMILES CC1=CC(=C(C(=C1)O)C)C

Spécifications

Melting Point 93°C
Color Blanc-jaune
Boiling Point 233°C
Quantity 25 g
UN Number 2430
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2,3,5-Trimethylphenol
EINECSNumber (3)-0540&(4)-0057&(9)-0899
TSCA Yes