missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, 98+%

Catalog No. AAA1074514
Change view
Click to view available options
Quantity:
25 g
100 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1074514 25 g
AAA1074522 100 g
AAA107450B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1074514 Supplier Thermo Scientific Chemicals Supplier No. A1074514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 697-82-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 697-82-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002228
InChI Key OGRAOKJKVGDSFR-UHFFFAOYSA-N
Synonym isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
PubChem CID 12769
ChEBI CHEBI:38570
IUPAC Name 2,3,5-trimethylphenol
SMILES CC1=CC(=C(C(=C1)O)C)C

Specifications

Melting Point 92°C to 95°C
Boiling Point 230°C to 231°C
Odor Phenol-like
Quantity 25 g
UN Number UN2430
Beilstein 2042210
Formula Weight 136.2
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Trimethylphenol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 211-806-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only