missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylfuran, 99%

Catalog No. p-7077484
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454470010 1 g
AC454470050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454470010 Supplier Thermo Scientific Chemicals Supplier No. 454470010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10504-04-8 | C7H10O | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10504-04-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD03931179
InChI Key NJXZFRUNHWKHEC-UHFFFAOYSA-N
Synonym furan, 2,3,5-trimethyl, 2,3,5-trimethyl-furan, furan,2,3,5-trimethyl
PubChem CID 517859
IUPAC Name 2,3,5-trimethylfuran
SMILES CC1=CC(C)=C(C)O1

Specifications

Density 0.8900g/mL
Boiling Point 121.0°C to 122.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.449
Quantity 1 g
Specific Gravity 0.89
Formula Weight 110.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylfuran
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H StatementHighly flammable liquid and vapour.Harmful if swallowed.Harmful in contact with skin.Causes skin irritation.Causes serious eye irritation.Harmful if inhaled.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only