missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trimethylfuran, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.8900g/mL |
| Boiling Point | 121.0°C to 122.0°C |
| Flash Point | 20°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Refractive Index | 1.449 |
| Quantity | 5 g |
| Specific Gravity | 0.89 |
| Formula Weight | 110.16 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H StatementHighly flammable liquid and vapour.Harmful if swallowed.Harmful in contact with skin.Causes skin irritation.Causes serious eye irritation.Harmful if inhaled. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only