missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triiodobenzoic Acid 98.0+%, TCI America™
$78.29 - $227.41
Chemical Identifiers
| CAS | 88-82-4 |
|---|---|
| Molecular Formula | C7H3I3O2 |
| Molecular Weight (g/mol) | 499.81 |
| MDL Number | MFCD00002420 |
| InChI Key | ZMZGFLUUZLELNE-UHFFFAOYSA-N |
| Synonym | floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova |
| PubChem CID | 6948 |
| ChEBI | CHEBI:73175 |
| IUPAC Name | 2,3,5-triiodobenzoic acid |
| SMILES | OC(=O)C1=CC(I)=CC(I)=C1I |
Chemical Identifiers
| 88-82-4 | |
| 499.81 | |
| ZMZGFLUUZLELNE-UHFFFAOYSA-N | |
| 6948 | |
| 2,3,5-triiodobenzoic acid |
| C7H3I3O2 | |
| MFCD00002420 | |
| floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova | |
| CHEBI:73175 | |
| OC(=O)C1=CC(I)=CC(I)=C1I |
Specifications
| 88-82-4 | |
| C7H3I3O2 | |
| 5 g | |
| ZMZGFLUUZLELNE-UHFFFAOYSA-N | |
| 2,3,5-triiodobenzoic acid | |
| 6948 | |
| 499.81 | |
| Crystalline Powder |
| 231°C | |
| MFCD00002420 | |
| floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova | |
| OC(=O)C1=CC(I)=CC(I)=C1I | |
| 499.81 | |
| CHEBI:73175 | |
| ≥98.0% (HPLC,T) | |
| 2,3,5-Triiodobenzoic Acid |
Safety and Handling
RTECSNumber : DH9275000
TSCA : Yes