missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triiodobenzoic Acid 98.0+%, TCI America™

Catalog No. T04515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04515G 5 g
T045125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04515G Supplier TCI America Supplier No. T04515G

Chemical Identifiers

CAS 88-82-4
Molecular Formula C7H3I3O2
Molecular Weight (g/mol) 499.81
MDL Number MFCD00002420
InChI Key ZMZGFLUUZLELNE-UHFFFAOYSA-N
Synonym floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
PubChem CID 6948
ChEBI CHEBI:73175
IUPAC Name 2,3,5-triiodobenzoic acid
SMILES OC(=O)C1=CC(I)=CC(I)=C1I

Specifications

Melting Point 231°C
Quantity 5 g
Formula Weight 499.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5-Triiodobenzoic Acid
RTECSNumber DH9275000
TSCA Yes