missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Triiodobenzoic acid, 98+%

Catalog No. AAL0267922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0267922 100 g
AAL0267906 5 g
AAL0267914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0267922 Supplier Thermo Scientific Chemicals Supplier No. L0267922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88-82-4 | C7H3I3O2 | 499.81 g/mol

2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.

Chemical Identifiers

CAS 88-82-4
Molecular Formula C7H3I3O2
Molecular Weight (g/mol) 499.81
MDL Number MFCD00002420
InChI Key ZMZGFLUUZLELNE-UHFFFAOYSA-N
Synonym floraltone, tiba, triiodobenzoic acid, johnkolor, benzoic acid, 2,3,5-triiodo, 2,3,5-tiba, caswell no. 890a, triiodobenzoic acid van, regin 8, kyselina 2,3,5-trijodbenzoova
PubChem CID 6948
ChEBI CHEBI:73175
IUPAC Name 2,3,5-triiodobenzoic acid
SMILES OC(=O)C1=CC(I)=CC(I)=C1I

Specifications

Melting Point 220°C to 222°C
Quantity 100 g
Beilstein 1955088
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 499.81
Percent Purity ≥98%
Chemical Name or Material 2,3,5-Triiodobenzoic acid
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-859-6
RTECSNumber DH9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only