missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triiodobenzoic acid, 98+%
Description
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
Specifications
Specifications
| Melting Point | 220°C to 222°C |
| Quantity | 5 g |
| Beilstein | 1955088 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 499.81 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2,3,5-Triiodobenzoic acid |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 201-859-6 |
| RTECSNumber | DH9275000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only