missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichloropyridine 98.0+%, TCI America™

Supplier:  TCI America T1479250G

 View more versions of this product

Catalog No. T1479250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3,5-Trichloropyridine
47°C
220°C
MFCD00043007
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
C1=C(C=NC(=C1Cl)Cl)Cl
182.428
182.43
Crystalline Powder
16063-70-0
White-Yellow
C5H2Cl3N
250 g
CNLIIAKAAMFCJG-UHFFFAOYSA-N
2,3,5-trichloropyridine
27666
≥98.0% (GC)

Chemical Identifiers

16063-70-0
182.428
CNLIIAKAAMFCJG-UHFFFAOYSA-N
27666
C1=C(C=NC(=C1Cl)Cl)Cl
C5H2Cl3N
MFCD00043007
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
2,3,5-trichloropyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5258

RTECSNumber : UU0525000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions