missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trichloropyridine 98.0+%, TCI America™
$100.11 - $598.72
Chemical Identifiers
| CAS | 16063-70-0 |
|---|---|
| Molecular Formula | C5H2Cl3N |
| Molecular Weight (g/mol) | 182.428 |
| MDL Number | MFCD00043007 |
| InChI Key | CNLIIAKAAMFCJG-UHFFFAOYSA-N |
| Synonym | pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 |
| PubChem CID | 27666 |
| IUPAC Name | 2,3,5-trichloropyridine |
| SMILES | C1=C(C=NC(=C1Cl)Cl)Cl |
Chemical Identifiers
| 16063-70-0 | |
| 182.428 | |
| CNLIIAKAAMFCJG-UHFFFAOYSA-N | |
| 27666 | |
| C1=C(C=NC(=C1Cl)Cl)Cl |
| C5H2Cl3N | |
| MFCD00043007 | |
| pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 | |
| 2,3,5-trichloropyridine |
Specifications
| 16063-70-0 | |
| White-Yellow | |
| C5H2Cl3N | |
| 25 g | |
| CNLIIAKAAMFCJG-UHFFFAOYSA-N | |
| 2,3,5-trichloropyridine | |
| 27666 | |
| ≥98.0% (GC) | |
| 2,3,5-Trichloropyridine |
| 47°C | |
| 220°C | |
| MFCD00043007 | |
| pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005 | |
| C1=C(C=NC(=C1Cl)Cl)Cl | |
| 182.428 | |
| 182.43 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-5258
RTECSNumber : UU0525000
TSCA : No