missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone, Thermo Scientific Chemicals

Catalog No. ac468640010
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AC468640010 1 g
AC468640050 5 g
2 options

Catalog No. AC468640010

Supplier: Thermo Scientific Chemicals 468640010

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55094-52-5
Molecular Formula C26H26O5
Molecular Weight (g/mol) 418.49
MDL Number MFCD08703966
InChI Key LDHBSABBBAUMCZ-UHFFFAOYNA-N
IUPAC Name 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one
SMILES O=C1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 53.0°C to 55.0°C
Quantity 1 g
Formula Weight 418.49
Chemical Name or Material 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only