missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-D-arabinofuranose

Catalog No. AAJ6679209
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6679209 10 g
AAJ6679206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6679209 Supplier Thermo Scientific Chemicals Supplier No. J6679209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37776-25-3 | C26H28O5 | 420.505 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37776-25-3
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00080813
InChI Key NAQUAXSCBJPECG-DYXQDRAXSA-N
Synonym 2,3,5-tri-o-benzyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-d-arabinofurannose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, d-ribose,2,3,5-tris-o-phenylmethyl, 2-o,3-o,5-o-tribenzyl-d-arabinofuranose, 2,3,5-tris-o-phenylmethyl-d-arabinofuranose, 2,3,5-tri-o-benzyl-alpha/beta-d-arabinofuranose, 3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 11058881
IUPAC Name (3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 10 g
Formula Weight 420.5
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-D-arabinofuranose
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only