missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

Catalog No. AAJ6625206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6625206 5 g
AAJ6625203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6625206 Supplier Thermo Scientific Chemicals Supplier No. J6625206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89361-52-4 | C26H28O5 | 420.505 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 89361-52-4
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD01864312
InChI Key NAQUAXSCBJPECG-VEYUFSJPSA-N
Synonym 2,3,5-tri-o-benzyl-b-d-ribofuranose, 2,3,5-tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 57369909
IUPAC Name (2R,3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in chloroform
Optical Rotation +43° (c=2 in Dioxane)
Formula Weight 420.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-ribofuranose
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only