missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

Catalog No. AAJ6685309
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6685309 10 g
AAJ6685306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6685309 Supplier Thermo Scientific Chemicals Supplier No. J6685309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60933-68-8 | C26H28O5 | 420.505 g/mol

An arabinofuranose derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An arabinofuranose derivative.

Solubility
Soluble in methanol

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 60933-68-8
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00047076
InChI Key NAQUAXSCBJPECG-FXSWLTOZSA-N
Synonym 2,3,5-tri-o-benzyl-beta-d-arabinofuranose, 2,3,5-tri-o-benzyl-b-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-arabinofuranose, 2r,3s,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
PubChem CID 11729473
IUPAC Name (2R,3S,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 10 g
Solubility Information Soluble in methanol
Formula Weight 420.5
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only