missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. D54311G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54311G 1 g
D5431200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54311G Supplier TCI America Supplier No. D54311G
Only null left

Chemical Identifiers

CAS 428820-95-5
Molecular Formula C12H15BN2O6
Molecular Weight (g/mol) 294.07
MDL Number MFCD11504965
InChI Key SFFDLKOCTJMCCJ-UHFFFAOYSA-N
Synonym 1,3-Dinitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dinitrophenylboronic Acid Pinacol Ester
PubChem CID 46739239
IUPAC Name 2-(3,5-dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 294.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane