accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNOONOO

CAS RN 428820-95-5

IUPAC Name: 2-(3,5-dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,5-dinitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 294.07
Molecular Formula: C12H15BN2O6
InChi Key: SFFDLKOCTJMCCJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results