missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%

Catalog No. AAH50333MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50333MD 250 mg
AAH5033303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50333MD Supplier Thermo Scientific Chemicals Supplier No. H50333MD

CAS: 62821-88-9 | C7H13N3 | 139.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62821-88-9
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.20
MDL Number MFCD04967722
InChI Key FXLOXQBWUPCCDL-UHFFFAOYSA-N
Synonym 2-3,5-dimethyl-1h-pyrazol-1-yl ethanamine, 2-3,5-dimethyl-pyrazol-1-yl-ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethan-1-amine, 2-3,5-dimethylpyrazol-1-yl ethanamine, 2-3,5-dimethylpyrazol-1-yl ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethylamine, 1-2-aminoethyl-3,5-dimethylpyrazole, 2-3,5-dimethylpyrazolyl ethylamine, acmc-209v2h, 2-3,5-dimethyl-1-pyrazolyl ethanamine
PubChem CID 1091687
SMILES CC1=NN(CCN)C(C)=C1

Specifications

Quantity 250 mg
UN Number UN2735
Sensitivity Air and moisture sensitive
IUPAC Name 2-(3,5-dimethyl-1H-pyrazol-1-yl)ethan-1-amine
Formula Weight 139.2
Percent Purity 97%
Chemical Name or Material 2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only