CAS RN 62821-88-9
IUPAC Name:
2-(3,5-dimethyl-1H-pyrazol-1-yl)ethan-1-amine
Synonyms:
2-(3,5-dimethylpyrazol-1-yl)ethanamine
Molecular Weight (g/mol):
139.2
Molecular Formula:
C7H13N3
InChi Key:
FXLOXQBWUPCCDL-UHFFFAOYSA-N
SMILES:
CC1=NN(CCN)C(C)=C1
1
–
1
of
1
results
1
Promotions Available
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%
CAS: 62821-88-9 Molecular Formula: C7H13N3 Molecular Weight (g/mol): 139.20 MDL Number: MFCD04967722 InChI Key: FXLOXQBWUPCCDL-UHFFFAOYSA-N Synonym: 2-3,5-dimethyl-1h-pyrazol-1-yl ethanamine,2-3,5-dimethyl-pyrazol-1-yl-ethylamine,2-3,5-dimethyl-1h-pyrazol-1-yl ethan-1-amine,2-3,5-dimethylpyrazol-1-yl ethanamine,2-3,5-dimethylpyrazol-1-yl ethylamine,2-3,5-dimethyl-1h-pyrazol-1-yl ethylamine,1-2-aminoethyl-3,5-dimethylpyrazole,2-3,5-dimethylpyrazolyl ethylamine,acmc-209v2h,2-3,5-dimethyl-1-pyrazolyl ethanamine PubChem CID: 1091687 SMILES: CC1=NN(CCN)C(C)=C1