missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. D50175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50175G 5 g
D50171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50175G Supplier TCI America Supplier No. D50175G

Chemical Identifiers

CAS 863868-36-4
Molecular Formula C12H15BF2O2
Molecular Weight (g/mol) 240.06
MDL Number MFCD11504963
InChI Key NBMGRDOMOTUSOT-UHFFFAOYSA-N
Synonym 3,5-Difluorophenylboronic Acid Pinacol Ester, 1,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
PubChem CID 46739301
IUPAC Name 2-(3,5-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=CC(F)=C1

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane