missing translation for 'onlineSavingsMsg'
Learn More

2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. D45755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45755G 5 g
D45751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45755G Supplier TCI America Supplier No. D45755G

Chemical Identifiers

CAS 68716-51-8
Molecular Formula C12H15BCl2O2
Molecular Weight (g/mol) 272.96
MDL Number MFCD05863906
InChI Key HKSNHZBPIBSVGI-UHFFFAOYSA-N
Synonym 1,3-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dichlorophenylboronic Acid Pinacol Ester
PubChem CID 2758200
IUPAC Name 2-(3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Cl)Cl

Specifications

Melting Point 53°C
Color White
Quantity 5 g
Formula Weight 272.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane