accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CClCl

CAS RN 68716-51-8

IUPAC Name: 2-(3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,5-dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 272.96
Molecular Formula: C12H15BCl2O2
InChi Key: HKSNHZBPIBSVGI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(Cl)=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results