missing translation for 'onlineSavingsMsg'
Learn More

2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. B50955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50955G 5 g
B50951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50955G Supplier TCI America Supplier No. B50955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69807-91-6
Molecular Formula C14H15BF6O2
Molecular Weight (g/mol) 340.07
MDL Number MFCD12405516
InChI Key GGMXSSNJKVWXMD-UHFFFAOYSA-N
Synonym 3,5-Bis(trifluoromethyl)phenylboronic Acid Pinacol Ester, 1,3-Bis(trifluoromethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
PubChem CID 9798005
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 67°C
Color White
Quantity 5 g
Formula Weight 340.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-[3,5-Bis(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane