missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetramethylpyrazine, Analytical Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012491
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012491 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012491 Supplier Merck Emd Millipore Supplier No. 95162100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,3,5,6-Tetramethylpyrazine is a flavor compound belonging to the class of pyrazines. It is found to be a main active ingredient of the traditional Chinese medicine, Chuanxiong and Ma-Huang herbs, used to treat asthma.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 1124-11-4
Physical Form Neat
Linear Formula C8H12N2
MDL Number MFCD00006146
Melting Point 77°C to 80°C; 85°C to 87°C
Boiling Point 190°C
Quantity 100 mg
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less