missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzotrifluoride 97.0+%, TCI America™

Catalog No. T18545G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18545G Supplier TCI America Supplier No. T18545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 651-80-9
Molecular Formula C7HF7
Molecular Weight (g/mol) 218.074
MDL Number MFCD00012385
InChI Key ZVPAJILXQHMKMT-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorobenzotrifluoride, 1,2,4,5-tetrafluoro-3-trifluoromethyl benzene, benzene, 1,2,4,5-tetrafluoro-3-trifluoromethyl, toluene, .alpha.,.alpha.,.alpha.,2,3,5,6-heptafluoro, 4h-heptafluorotoluene, 1-trifluoromethyl-2,3,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorobenzotrifluoride_x000d_, alpha,alpha,alpha,2,3,5,6-heptafluorotoluene, 1,2,4,5-tetrafluoro-3-trifluoromethyl benzene #
PubChem CID 136465
IUPAC Name 1,2,4,5-tetrafluoro-3-(trifluoromethyl)benzene
SMILES C1=C(C(=C(C(=C1F)F)C(F)(F)F)F)F

Specifications

Color Colorless
Boiling Point 112°C
Quantity 5 g
UN Number 1993
Formula Weight 218.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator