missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoroanisole 97.0+%, TCI America™

Catalog No. T10415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10415G Supplier TCI America Supplier No. T10415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 2324-98-3
Color Colorless
Boiling Point 145°C
Molecular Formula C7H4F4O
Quantity 5 g
MDL Number MFCD00000311
UN Number 1993
Synonym 2,3,5,6-tetrafluoroanisole, 2,3,5,6-tetrafluoromethoxy benzene, anisole, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoromethoxybenzene, methyl 2,3,5,6-tetrafluorophenyl ether, benzene, 1,2,4,5-tetrafluoro-3-methoxy, 1,2,4,5-tetrafluoro-3-methoxy-benzene, pubchem10114, acmc-1clho, 2,3,5,6-tetrafluoro-anisole
InChI Key AXCOCGJDERQVDK-UHFFFAOYSA-N
SMILES COC1=C(F)C(F)=CC(F)=C1F
IUPAC Name 1,2,4,5-tetrafluoro-3-methoxybenzene
Molecular Weight (g/mol) 180.10
PubChem CID 75351
Formula Weight 180.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluoroanisole
Show More Show Less