missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzenethiol 97.0+%, TCI America™

Catalog No. T15421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15421G 1 g
T15425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15421G Supplier TCI America Supplier No. T15421G

Chemical Identifiers

CAS 651-84-3
Molecular Formula C7HF7S
Molecular Weight (g/mol) 250.13
MDL Number MFCD00191594
InChI Key BXMOMKVOHOSVJH-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-trifluoromethyl benzenethiol, 4-trifluoromethyl-2,3,5,6-tetrafluorothiophenol, 4-trifluoromethyl tetrafluorothiophenol, 2,3,5,6-tetrafluoro-4-trifluoromethyl benzene-1-thiol, benzenethiol,2,3,5,6-tetrafluoro-4-trifluoromethyl, acmc-209npo, tetrafluoro-4-trifluoromethyl thiophenol, 2,3,5,6-tetra-fluoro-4-trifluoromethylbenzenethiol, 4-mercapto-alpha,alpha,alpha,2,3,5,6-heptafluorotoluene
PubChem CID 619538
IUPAC Name 2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene-1-thiol
SMILES FC1=C(F)C(=C(F)C(F)=C1S)C(F)(F)F

Specifications

Color Yellow
Boiling Point 169°C
Quantity 1 g
Formula Weight 250.13
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzenethiol