missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5,6-Tetrachloropyridine 98.0+%, TCI America™
$117.43 - $365.35
Chemical Identifiers
| CAS | 2402-79-1 |
|---|---|
| Molecular Formula | C5HCl4N |
| Molecular Weight (g/mol) | 216.87 |
| MDL Number | MFCD00092428 |
| InChI Key | FATBKZJZAHWCSL-UHFFFAOYSA-N |
| Synonym | pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine |
| PubChem CID | 16990 |
| IUPAC Name | 2,3,5,6-tetrachloropyridine |
| SMILES | ClC1=CC(Cl)=C(Cl)N=C1Cl |
Chemical Identifiers
| 2402-79-1 | |
| 216.87 | |
| FATBKZJZAHWCSL-UHFFFAOYSA-N | |
| 16990 | |
| ClC1=CC(Cl)=C(Cl)N=C1Cl |
| C5HCl4N | |
| MFCD00092428 | |
| pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine | |
| 2,3,5,6-tetrachloropyridine |
Specifications
| 2402-79-1 | |
| White | |
| C5HCl4N | |
| 5 g | |
| FATBKZJZAHWCSL-UHFFFAOYSA-N | |
| 2,3,5,6-tetrachloropyridine | |
| 16990 | |
| ≥98.0% (GC) | |
| 2,3,5,6-Tetrachloropyridine |
| 93°C | |
| 251°C | |
| MFCD00092428 | |
| pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine | |
| ClC1=CC(Cl)=C(Cl)N=C1Cl | |
| 216.87 | |
| 216.87 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UT8225000
TSCA : Yes