missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrachlorophenol, MilliporeSigma™ Supelco™

Catalog No. 111019513
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019513 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019513 Supplier Merck Emd Millipore Supplier No. 442284

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,3,5,6-Tetrachlorophenol belongs to the class of substituted phenol compounds, which are widely used in industrial applications which include synthesis of pesticides, dyes, drugs, plastics, etc. They are found in environmental samples as pollutant due to their extensive usage.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 935-95-5
Physical Form Neat
Linear Formula C6H2 Cl4 O
MDL Number MFCD00044671
Type Organic Pollutant Standard
Quantity 50 mg
Formula Weight 231.89
Grade Analytical Standard