missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrabromo-p-xylene 98.0+%, TCI America™

Catalog No. T32021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32021G 1 g
T32025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32021G Supplier TCI America Supplier No. T32021G
Only null left

Chemical Identifiers

CAS 23488-38-2
Molecular Formula C8H6Br4
Molecular Weight (g/mol) 421.752
MDL Number MFCD00010353
InChI Key RXKOKVQKECXYOT-UHFFFAOYSA-N
Synonym 1,2,4,5-Tetrabromo-3,6-dimethylbenzene
PubChem CID 31952
IUPAC Name 1,2,4,5-tetrabromo-3,6-dimethylbenzene
SMILES CC1=C(C(=C(C(=C1Br)Br)C)Br)Br

Specifications

Melting Point 256°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 421.75
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3,5,6-Tetrabromo-p-xylene
RTECSNumber DB9072000
TSCA Yes