missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trimethoxybenzoic Acid 98.0+%, TCI America™
Supplier: TCI America T11165G
Specifications
| 2,3,4-Trimethoxybenzoic Acid | |
| 100°C | |
| C10H12O5 | |
| 5 g | |
| HZNQSWJZTWOTKM-UHFFFAOYSA-N | |
| 2,3,4-trimethoxybenzoic acid | |
| 11308 | |
| ≥98.0% (GC,T) |
| 573-11-5 | |
| White | |
| MFCD00002433 | |
| benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid | |
| COC1=C(C(=C(C=C1)C(=O)O)OC)OC | |
| 212.201 | |
| 212.20 | |
| Crystalline Powder |
Chemical Identifiers
| 573-11-5 | |
| 212.201 | |
| HZNQSWJZTWOTKM-UHFFFAOYSA-N | |
| 11308 | |
| COC1=C(C(=C(C=C1)C(=O)O)OC)OC |
| C10H12O5 | |
| MFCD00002433 | |
| benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid | |
| 2,3,4-trimethoxybenzoic acid |
Safety and Handling
TSCA : No