Learn More
2,3,4-Trimethoxybenzoic acid, 98+%
CAS: 573-11-5 | C10H12O5 | 212.201 g/mol
Supplier: Thermo Scientific Chemicals A1427830
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,3,4-Trimethoxybenzoic acid | |
| 99°C to 102°C | |
| C10H12O5 | |
| 250 g | |
| benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid | |
| COC1=C(C(=C(C=C1)C(=O)O)OC)OC | |
| 212.201 | |
| 212.2 |
| 573-11-5 | |
| Odorless | |
| MFCD00002433 | |
| 2696073 | |
| HZNQSWJZTWOTKM-UHFFFAOYSA-N | |
| 2,3,4-trimethoxybenzoic acid | |
| 11308 | |
| ≥98% |
Chemical Identifiers
| 573-11-5 | |
| 212.201 | |
| HZNQSWJZTWOTKM-UHFFFAOYSA-N | |
| 11308 | |
| COC1=C(C(=C(C=C1)C(=O)O)OC)OC |
| C10H12O5 | |
| MFCD00002433 | |
| benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid | |
| 2,3,4-trimethoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-350-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only