missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trimethoxybenzoic acid, 98+%

Catalog No. AAA1427809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1427809 10 g
AAA1427818 50 g
AAA1427830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1427809 Supplier Thermo Scientific Chemicals Supplier No. A1427809
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 573-11-5 | C10H12O5 | 212.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 573-11-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002433
InChI Key HZNQSWJZTWOTKM-UHFFFAOYSA-N
Synonym benzoic acid, 2,3,4-trimethoxy, 2,3,4-trimethoxybenzoicacid, 2,3,4-trimethoxy-benzoic acid, trimethoxybenzoic acid, benzoic acid, trimethoxy, acmc-209lx4, ksc265s3r, 2,3,4-trimethoxy benzoic acid, 2,3,4-trimethoxybenzoic acid
PubChem CID 11308
IUPAC Name 2,3,4-trimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)C(=O)O)OC)OC

Specifications

Melting Point 99°C to 102°C
Odor Odorless
Quantity 10 g
Beilstein 2696073
Formula Weight 212.2
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-350-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only