missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trimethoxyacetophenone 97.0+%, TCI America™

Catalog No. T15175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15175G Supplier TCI America Supplier No. T15175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13909-73-4
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00008726
InChI Key PKNAATJMQOUREZ-UHFFFAOYSA-N
Synonym 2',3',4'-trimethoxyacetophenone, 2,3,4-trimethoxyacetophenone, 1-2,3,4-trimethoxyphenyl ethanone, ethanone, 1-2,3,4-trimethoxyphenyl, unii-f3jc7zp11a, f3jc7zp11a, ethanone,1-2,3,4-trimethoxyphenyl, 2',3',4' trimethoxyacetophenone, 1-acetyl-2,3,4-trimethoxybenzene, 1-2,3,4-trimethoxyphenyl ethan-1-one
PubChem CID 83810
IUPAC Name 1-(2,3,4-trimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)OC)OC)OC

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,3′,4′-Trimethoxyacetophenone
EINECSNumber (3)-1255
RTECSNumber AN0531000
TSCA No