CAS RN 13909-73-4
IUPAC Name:
1-(2,3,4-trimethoxyphenyl)ethan-1-one
Synonyms:
2,3,4-trimethoxyacetophenone
Molecular Weight (g/mol):
210.23
Molecular Formula:
C11H14O4
InChi Key:
PKNAATJMQOUREZ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(C)=O)C(OC)=C1OC
1
–
1
of
1
results
1
2',3',4'-Trimethoxyacetophenone 97.0+%, TCI America™
CAS: 13909-73-4 Molecular Formula: C11H14O4 Molecular Weight (g/mol): 210.229 MDL Number: MFCD00008726 InChI Key: PKNAATJMQOUREZ-UHFFFAOYSA-N Synonym: 2',3',4'-trimethoxyacetophenone,2,3,4-trimethoxyacetophenone,1-2,3,4-trimethoxyphenyl ethanone,ethanone, 1-2,3,4-trimethoxyphenyl,unii-f3jc7zp11a,f3jc7zp11a,ethanone,1-2,3,4-trimethoxyphenyl,2',3',4' trimethoxyacetophenone,1-acetyl-2,3,4-trimethoxybenzene,1-2,3,4-trimethoxyphenyl ethan-1-one PubChem CID: 83810 IUPAC Name: 1-(2,3,4-trimethoxyphenyl)ethanone SMILES: CC(=O)C1=C(C(=C(C=C1)OC)OC)OC