missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzophenone 98.0+%, TCI America™

Catalog No. T099625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T099625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T099625G Supplier TCI America Supplier No. T099625G

Chemical Identifiers

CAS 1143-72-2
Molecular Formula C13H10O4
Molecular Weight (g/mol) 230.219
MDL Number MFCD00009996
InChI Key HTQNYBBTZSBWKL-UHFFFAOYSA-N
Synonym 2,3,4-trihydroxybenzophenone, phenyl 2,3,4-trihydroxyphenyl methanone, alizarin yellow a, gallobenzophenone, alizarine yellow a, methanone, phenyl 2,3,4-trihydroxyphenyl, unii-8fbx2q800v, 2,3,4-trihydroxybenzophenon, chembl1527, 2,3,4-trihydroxbenzophenone
PubChem CID 70837
IUPAC Name phenyl-(2,3,4-trihydroxyphenyl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C(=C(C=C2)O)O)O

Specifications

Melting Point 141°C
Color Yellow
Quantity 25 g
Formula Weight 230.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trihydroxybenzophenone
EINECSNumber (4)-1460
TSCA Yes