missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T25745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25745G Supplier TCI America Supplier No. T25745G
Only null left

Chemical Identifiers

CAS 226396-32-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD01863168
InChI Key CLGIPVVEERQWSQ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorophenyl boronic acid, 2,3,4-trifluorobenzeneboronic acid, 2,3,4-trifluorphenylboronic acid, 4,5,6-trifluorophenylboronic acid, boronic acid, 2,3,4-trifluorophenyl, pubchem1887, acmc-1ckzl, trifluorophenylboronic acid, ksc201q5d, jalor-chem i01-8460
PubChem CID 2783280
IUPAC Name (2,3,4-trifluorophenyl)boronic acid
SMILES B(C1=C(C(=C(C=C1)F)F)F)(O)O

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.90
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trifluorophenylboronic Acid (contains varying amounts of Anhydride)