missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluoronitrobenzene 98.0+%, TCI America™

Catalog No. T15395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15395G 5 g
T153925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15395G Supplier TCI America Supplier No. T15395G
Only null left

Chemical Identifiers

CAS 771-69-7
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00041546
InChI Key ARCACZWMYGILNI-UHFFFAOYSA-N
Synonym 2,3,4-trifluoronitrobenzene, 2,3,4-trifluoro-1-nitrobenzene, 4-nitro-1,2,3-trifluorobenzene, trifluoronitrobenzene, benzene, 1,2,3-trifluoro-4-nitro, 2,3,4-trifluornitrobenzene, 1,2,3-trifluoro-4-nitro-benzene, 1-nitro-2,3,4-trifluorobenzene, pubchem2287, benzene, trifluoronitro
PubChem CID 69871
IUPAC Name 1,2,3-trifluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C(F)=C1F

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 177.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trifluoronitrobenzene