missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trifluorobenzyl Bromide 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 3265 |
| Synonym | 2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide |
| Formula Weight | 225.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3,4-Trifluorobenzyl Bromide |
Safety and Handling
| TSCA | No |