missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trifluorobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America T18685G
Specifications
| 2,3,4-Trifluorobenzoyl Chloride | |
| Yellow | |
| MFCD00075078 | |
| 3265 | |
| NXRQXCFBZGIRGN-UHFFFAOYSA-N | |
| 2,3,4-trifluorobenzoyl chloride | |
| 2734022 | |
| ≥98.0% (T) |
| 157373-08-5 | |
| C7H2ClF3O | |
| 5 g | |
| 2,3,4-trifluorobenzoylchloride, 2,3,4-trifluoro-benzoyl chloride, benzoyl chloride, 2,3,4-trifluoro, benzoyl chloride, 2,3,4-trifluoro-9ci, trifluorobenzoylchloride, trifluorobenzoyl chloride, intermediates-zcf02190, acmc-209dg2, 2,3,4-trifluorobenzoyl chloride | |
| C1=CC(=C(C(=C1C(=O)Cl)F)F)F | |
| 194.537 | |
| 194.54 | |
| Liquid |
Chemical Identifiers
| 157373-08-5 | |
| 194.537 | |
| NXRQXCFBZGIRGN-UHFFFAOYSA-N | |
| 2734022 | |
| C1=CC(=C(C(=C1C(=O)Cl)F)F)F |
| C7H2ClF3O | |
| MFCD00075078 | |
| 2,3,4-trifluorobenzoylchloride, 2,3,4-trifluoro-benzoyl chloride, benzoyl chloride, 2,3,4-trifluoro, benzoyl chloride, 2,3,4-trifluoro-9ci, trifluorobenzoylchloride, trifluorobenzoyl chloride, intermediates-zcf02190, acmc-209dg2, 2,3,4-trifluorobenzoyl chloride | |
| 2,3,4-trifluorobenzoyl chloride |
Safety and Handling
TSCA : No