missing translation for 'onlineSavingsMsg'
Learn More

2',3,4-Trifluoro-4″-propyl-1,1':4',1″-terphenyl 98.0+%, TCI America™

Catalog No. T34195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34195G 5 g
T34191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34195G Supplier TCI America Supplier No. T34195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 248936-60-9
Molecular Formula C21H17F3
Molecular Weight (g/mol) 326.36
MDL Number MFCD20488039
InChI Key MNDRQVSIWFDBFZ-UHFFFAOYSA-N
Synonym 2′,3,4-Trifluoro-4′′-propyl-p-terphenyl
PubChem CID 72376517
IUPAC Name 2,3',4'-trifluoro-4-(4-propylphenyl)-1,1'-biphenyl
SMILES CCCC1=CC=C(C=C1)C1=CC(F)=C(C=C1)C1=CC(F)=C(F)C=C1

Specifications

Color White
Quantity 5 g
Formula Weight 326.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3,4-Trifluoro-4′′-propyl-1,1′:4′,1′′-terphenyl