missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. d29635g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29635G Supplier TCI America Supplier No. D29635G

Chemical Identifiers

CAS 42880-07-9
Molecular Formula C15H11Cl6N3O2
Molecular Weight (g/mol) 477.972
MDL Number MFCD04038154
InChI Key ZJRNXDIVAGHETA-UHFFFAOYSA-N
PubChem CID 53384523
IUPAC Name 2-[2-(3,4-dimethoxyphenyl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES COC1=C(C=C(C=C1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl)OC

Specifications

Melting Point 190°C
Color Yellow
Quantity 5 g
Formula Weight 477.97
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,4-Dimethoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine