missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxyphenyl)ethanol 98.0+%, TCI America™

Catalog No. p-7128099
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41075G 5 g
D410725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41075G Supplier TCI America Supplier No. D41075G

Chemical Identifiers

CAS 7417-21-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00002894
InChI Key SRQAJMUHZROVHW-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene
PubChem CID 81911
IUPAC Name 2-(3,4-dimethoxyphenyl)ethan-1-ol
SMILES COC1=CC=C(CCO)C=C1OC

Specifications

Melting Point 46°C
Boiling Point 174°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)ethanol