missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxyphenyl)ethanol, 98%

Catalog No. AAH2694109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2694109 10 g
AAH2694118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2694109 Supplier Thermo Scientific Chemicals Supplier No. H2694109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7417-21-2 | C10H14O3 | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7417-21-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00002894
InChI Key SRQAJMUHZROVHW-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene
PubChem CID 81911
SMILES COC1=CC=C(CCO)C=C1OC

Specifications

Melting Point 45°C to 49°C
Boiling Point 172°C to 174°C (17 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
IUPAC Name 2-(3,4-dimethoxyphenyl)ethanol
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)ethanol
EINECSNumber 231-032-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only