accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Chromatography Standards
Organic Standards
06801642
Structure Search
Print
2,3,4,6-Tetrachlorophenol, Restek
Catalog No.
06801642
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
06801642
1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
06801642
Supplier
Chromatographic
Supplier No.
RK 31402
Sign In
or
Register
to check your price and availability.
Quantity
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
58-90-2
Molecular Formula
C6H2Cl4O
Molecular Weight (g/mol)
231.881
InChI Key
VGVRPFIJEJYOFN-UHFFFAOYSA-N
PubChem CID
6028
IUPAC Name
2,3,4,6-tetrachlorophenol
SMILES
C1=C(C(=C(C(=C1Cl)Cl)Cl)O)Cl
Specifications
Quantity
1 mL
Chemical Name or Material
2,3,4,6-Tetrachlorophenol
Product Suggestions
Documents