missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetrachlorophenol, MilliporeSigma™ Supelco™

Catalog No. 111019694
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019694 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019694 Supplier Merck Emd Millipore Supplier No. 442282
Only null left

2,3,4,6-Tetrachlorophenol belongs to the class of substituted phenol compounds, which are widely used in industrial applications such as synthesis of pesticides, dyes, drugs, plastics, etc. Due to their extensive usage, they are commonly found in environmental samples as pollutants.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 58-90-2
Physical Form Neat
Linear Formula C6H2 Cl4 O
MDL Number MFCD00019979
Type Organic Pollutant Standard
Quantity 100 mg
Formula Weight 231.89
Grade Analytical Standard